TISSUE-TROL™ Control Slides Argentaffin
Leverandør: SIGMA ALDRICH MICROSCOPY
TTR013-25EA
TTR013-25EAEA 2570 NOK
TTR013-25EA
TISSUE-TROL™ Control Slides Argentaffin
Objektglass Microscopy Control Slides
Histology slides with paraffin embedded section of human intestine containing argentaffin granules. Suitable for use with Fontana-Masson stain kit (Procedure No. HT200).
Leveringsinformasjon: Box contains 24 unstained slides plus one reference slide.