Control slide, Melanin TISSUE-TROL™, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
TTR014-25EA
TTR014-25EAEA 2570 NOK
TTR014-25EA
Control slide, Melanin TISSUE-TROL™, Sigma-Aldrich®
Objektglass Microscopy Control Slides
Histology slides with paraffin embedded section of human skin containing melanin.
Suitable for use with Fontana-Masson Stain Kit (Procedure No. HT200).
Bestill nå
SPESIFIKASJONER
Arter