Polyethylene glycol 8,000, crystalline powder, J.T.Baker®

Leverandør: Avantor

Synonymer: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000

U222-01EA 0 NOK
U222-01
Polyethylene glycol 8,000, crystalline powder, J.T.Baker®
Polyethylene glycol 8,000

Powder.

Formel: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Kokepunkt: 250 °C (1013 hPa)
Tetthet: 1,125 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01779615
CAS-nummer: 25322-68-3
EINECS: 500-038-2
Merck Indeks: 12,07729

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance Passes Test
Viscosity, cSt 470 - 900
Average Molecular Weight 7000 - 9000

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR