Face shield visor, 701.00
Leverandør: UNIVET
2200014
UNVT2200014EA 825 NOK
UNVT2200014
Face shield visor, 701.00
Ansiktsskjermer
The 701.00 face shield provides protection while ensuring comfort with a soft latex-free sponge on the forehead.
  • Anti-fog acetate visor
  • Soft latex-free sponge on the forehead
  • Adjustable elastic head band
  • Visor inclines up to 90°
  • Interchangeable visor
  • Face shield suitable for use in combination with prescription glasses and protective masks
The thermo-formed acetate screen maximizes vision due to anti-fog characteristics. The visor can be tilted up to an angle of 90 degrees, and is easily replaceable. The 701.00 visor is compatible with respiratoryMore Product Information