Safety goggle, 601
Leverandør: UNIVET
601.00.77.00
UNVT601.00.77.00EA 1575 NOK
UNVT601.00.77.00
Safety goggle, 601
Vernebriller /Goggles
The breadth of the 601 range provides a superior versatility.
  • Wide field of vision for an high-tech goggle
  • The anatomic nose pad fits perfectly to all faces
  • Directly vented with foam
  • No colour distortion
  • Mirrored version
The various technologies of the ventilation system and polycarbonate lens make it suitable for wide range of sectors with high level of performance.
The adjustable headband and the soft frame effectively distribute the pressure on the face.
This goggle is suitable for mechanical work in good visibility conditions.