Face shields, 607
Leverandør: UNIVET
607.A7.00.00
UNVT607.A7.00.00EA 330 NOK
UNVT607.A7.00.00
Face shields, 607
Ansiktsskjermer
Visor made from clear injected PC.
  • Can be combined with headbands or helmets
  • Provides protection against electric arcs caused by short circuits
It provides protection against UV radiation, droplets and splashes, mechanical impacts, molten metals and hot solids.
Thickness: 2 mm
Sertifiseringer: Cat. III
Bestillingsinformasjon: The visor is supplied without headband or helmet. These need to be ordered separately.