Tubing expander, Upchruch Scientific®
Leverandør: UPCHURCH SCIENTIFIC
P-780
UPCHP-780EA 740 NOK
UPCHP-780
Tubing expander, Upchruch Scientific®
Kromatografiutstyr Kromatografireservedeler
Use this tubing expanding awl when working with barbed fittings. Sometimes the tubing can be too tight to fit over the barb properly. This awl can also be used with semi-rigid tubing, for those rare applications where this tubing needs to be paired with a barbed fitting. By expanding the int.Ø of the tubing, it will make it an easier process to insert the barbed fitting into place.