Burner, Teclu, with stopcock
Leverandør: USBECK Laborgeräte
1240
USBE1240EA 1725 NOK
USBE1240
Burner, Teclu, with stopcock
Termostater Gassbrennere
Burner for natural gas, adjustable. Maximum temperature of 1620 °C.