Safety burners with stopcock
Leverandør: USBECK Laborgeräte
1340 1341
451-1369EA 3500 NOK
451-1369 USBE1341
Safety burners with stopcock
Termostater Gassbrennere
This safety burner provides protection against uncontrolled escape of gas in laboratories. If the flame is extinguished, the gas supply is interrupted and no unburnt gas can escape.
  • Safety pilot flame
  • 90° stopcock
  • With air regulation and thermocouple