Table clamp
Leverandør: USBECK Laborgeräte
2051
USBE2051EA 1700 NOK
USBE2051
Table clamp
Klemmer Universalklemmer
Large table clamp made of powder-coated aluminium alloy.
  • For rods and square rods up to 26 mm Ø
  • For benches up to 55 mm