Dobbelmuffer, kuleformede
Leverandør: USBECK Laborgeräte
2360 2362 2363
USBE2360EA 255 NOK
USBE2360 USBE2362 USBE2363
Dobbelmuffer, kuleformede
Klemmer Kryssmuffer
Bossheads made of zinc die-cast, either natural, powder coated or nickel plated.
  • 35 mm Ø
  • For rods up to 13 mm Ø
  • Nickel plated hinged screws
Weight: 100 g