Clamps, Zinc Die Cast
Leverandør: USBECK Laborgeräte
2524 2534
USBE2524EA 435 NOK
USBE2524 USBE2534
Clamps, Zinc Die Cast
Klemmer Universalklemmer
These clamps are zinc diecasted, nickel plated with vinyl coated jaws. Rods are 10 mm in diameter.
Advantages of zinc diecasting:
Better energy efficiency during casting process thanks to a lower melting point, when compared to aluminum or malleable cast iron
High recycling proportion of at least 75% of raw material used
High stability also of thin-walled parts e.g. bossheads
Excellent cost-performance ratio of our basic assortment
Good corrosion resistance - protection through natural passivation (reaction of zinc with oxygen at the surface)
Corrosion resistance can be further enhanced byMore Product Information