Crucibles, stainless steel
Leverandør: USBECK Laborgeräte
5640 5641 5642 5643
USBE5640EA 330 NOK
USBE5640 USBE5641 USBE5642 USBE5643
Crucibles, stainless steel
Digler
Crucibles without lid made from stainless steel 18/10.
  • Highly resistant to chemicals and reagents
  • Withstand maximum temperature of 800 °C
  • Autoclavable
Bestillingsinformasjon: Lids need to be ordered separately.