Anti-JAM3 Rabbit Polyclonal Antibody

Leverandør: US Biological
029520
USBI029520EA 7400 NOK
USBI029520
Anti-JAM3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0. 05 ?g/ml

Type: Primary
Antigen: JAM3 (junctional adhesion molecule 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR