Anti-JAK3 Mouse Monoclonal Antibody [clone: 13B433]

Leverandør: US Biological
030355
USBI030355EA 7050 NOK
USBI030355
Anti-JAK3 Mouse Monoclonal Antibody [clone: 13B433]
Antistoffer
Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: Monoclonal
Clone: 13B433
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR