Anti-ADCY10 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
031626-FITC
USBI031626-FITCEA 10875 NOK
USBI031626-FITC
Anti-ADCY10 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: ADCY10 (adenylate cyclase 10 (soluble))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol ADCY10
Antigen-navn adenylate cyclase 10 (soluble)
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Flowcytometri Yes
Epitop internal
Aminosyretall 564 to 591
Skjema liquid
Rensing purified by Protein A chromatography
Lagringsbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR