Anti-ATF4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
032162-FITC
USBI032162-FITCEA 10875 NOK
USBI032162-FITC
Anti-ATF4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: FLISA1:1000 Dot blot 1:500
Do not freeze!

Type: Primary
Antigen: ATF4 (activating transcription factor 4)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR