Anti-FCGRT Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
035585-FITC
USBI035585-FITCEA 11100 NOK
USBI035585-FITC
Anti-FCGRT Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Primary
Antigen: FCGRT (Fc fragment of IgG, receptor, transporter, alpha)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR