Anti-HAT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
036434-FITC
USBI036434-FITCEA 10875 NOK
USBI036434-FITC
Anti-HAT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: HAT1 (histone acetyltransferase 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol HAT1
Environmentally Preferable
Antigen-navn Histone acetyltransferase 1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitop N-terminal
Aminosyretall 20 to 50
Skjema liquid
Rensing purified by Protein A chromatography
Lagringsbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR