Anti-IGFBP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: US Biological
036914-HRP
USBI036914-HRPEA 10875 NOK
USBI036914-HRP
Anti-IGFBP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: IGFBP4 (insulin-like growth factor binding protein 4)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol IGFBP4
Antigen-navn Insulin-Like Growth Factor Binding Protein 4
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering HRP (Horseradish Peroxidase)
Reaktivitet Human,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitop internal
Aminosyretall 103 to 132
Skjema liquid
Rensing purified by Protein A chromatography
Lagringsbuffer PBS, pH 7,2
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR