Anti-IGFBP4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverandør: US Biological
036914-PE
USBI036914-PEEA 10875 NOK
USBI036914-PE
Anti-IGFBP4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: IGFBP4 (insulin-like growth factor binding protein 4)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR