Anti-JAK3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: US Biological
037180-AP
USBI037180-APEA 10875 NOK
USBI037180-AP
Anti-JAK3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR