Anti-JAK3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: US Biological
037180-APC
USBI037180-APCEA 10875 NOK
USBI037180-APC
Anti-JAK3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol JAK3
Antigen-navn Janus kinase 3
Antistofftype Primary
Klonalitet Polyclonal
Konjugering APC (Allophycocyanin)
Reaktivitet Human,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitop N-terminal
Aminosyretall 132 to 161
Skjema liquid
Rensing purified by Protein A chromatography
Lagringsbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Lagringstemp Cool
Frakttemp. Ice
Uttestet applikasjon Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR