Anti-MCAF2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
038259-FITC
USBI038259-FITCEA 10875 NOK
USBI038259-FITC
Anti-MCAF2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: MCAF2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol MCAF2
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Human,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Flowcytometri Yes
Epitop internal
Aminosyretall 325 to 355
Skjema liquid
Rensing purified by Protein A chromatography
Lagringsbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Lagringstemp Cool
Frakttemp. Ice
Uttestet applikasjon Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR