Anti-MIIP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
038436-FITC
USBI038436-FITCEA 10875 NOK
USBI038436-FITC
Anti-MIIP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: Western Blot: 1:1000
Do not freeze!

Type: Primary
Antigen: MIIP (migration and invasion inhibitory protein)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR