Anti-MPI Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverandør: US Biological
038560-PE
USBI038560-PEEA 10875 NOK
USBI038560-PE
Anti-MPI Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MPI (mannose phosphate isomerase)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR