Anti-P-Glycoprotein Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
039557-BIOTIN
USBI039557-BIOTINEA 10875 NOK
USBI039557-BIOTIN
Anti-P-Glycoprotein Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: P-Glycoprotein
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigen-navn P-Glycoprotein
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitop internal
Aminosyretall 647 to 677
Skjema liquid
Rensing purified by Protein G chromatography
Lagringsbuffer PBS, pH 7,2
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR