Anti-SLC8A3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverandør: US Biological
041990-PE
USBI041990-PEEA 10875 NOK
USBI041990-PE
Anti-SLC8A3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: SLC8A3 (solute carrier family 8 (sodium/calcium exchanger), member 3)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR