Anti-STAR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
042371-FITC
USBI042371-FITCEA 10875 NOK
USBI042371-FITC
Anti-STAR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: STAR (steroidogenic acute regulatory protein)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR