Anti-STAR Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: US Biological
042371-HRP
USBI042371-HRPEA 10875 NOK
USBI042371-HRP
Anti-STAR Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: STAR (steroidogenic acute regulatory protein)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol STAR
Environmentally Preferable
Antigen-navn Steroidogenic Acute Regulatory Protein
Antistofftype Primary
Klonalitet Polyclonal
Konjugering HRP (Horseradish Peroxidase)
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitop internal
Aminosyretall 87 to 117
Skjema liquid
Rensing purified by Protein A chromatography
Lagringsbuffer PBS, pH 7,2
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR