Anti-TMCO5B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: US Biological
042919-AP
USBI042919-APEA 10875 NOK
USBI042919-AP
Anti-TMCO5B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TMCO5B (transmembrane and coiled-coil domains 5B, pseudogene)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR