Anti-TMCO5B Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverandør: US Biological
042919-PE
USBI042919-PEEA 10875 NOK
USBI042919-PE
Anti-TMCO5B Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TMCO5B (transmembrane and coiled-coil domains 5B, pseudogene)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR