Anti-USP7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: US Biological
043678-FITC
USBI043678-FITCEA 11100 NOK
USBI043678-FITC
Anti-USP7 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Primary
Antigen: USP7 (ubiquitin specific peptidase 7 (herpes virus-associated))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR