Anti-WAS Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
043802-BIOTIN
USBI043802-BIOTINEA 10875 NOK
USBI043802-BIOTIN
Anti-WAS Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: WAS (Wiskott-Aldrich syndrome)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR