Anti-MPI Mouse Monoclonal Antibody [clone: 13k85]

Leverandør: US Biological
143212
USBI143212EA 5175 NOK
USBI143212
Anti-MPI Mouse Monoclonal Antibody [clone: 13k85]
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:300

Type: Primary
Antigen: MPI (mannose phosphate isomerase)
Clonality: monoclonal
Clone: 13k85
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol MPI
Antigen-navn mannose phosphate isomerase
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon 13k85
Reaktivitet Human,Rat
Vert Mouse
Isotype IgG1
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing purified by immunoaffinity chromatography
Lagringsbuffer 0,1M Tris-glycine, pH 7,4, 150mM NaCl, 0,2% sodium azide, 50% glycerol.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR