Anti-SYP Mouse Monoclonal Antibody [clone: Syn-58]

Leverandør: US Biological
145243
USBI145243EA 7225 NOK
USBI145243
Anti-SYP Mouse Monoclonal Antibody [clone: Syn-58]
Antistoffer
Type: Primary
Antigen: SYP (synaptophysin)
Clonality: monoclonal
Clone: Syn-58
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR