Anti-NO66 Goat Polyclonal Antibody

Leverandør: US Biological
151191
USBI151191EA 9950 NOK
USBI151191
Anti-NO66 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:64000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: NO66
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR