Anti-ANK2 Mouse Monoclonal Antibody [clone: S105-17]

Leverandør: US Biological
167576
USBI167576EA 6325 NOK
USBI167576
Anti-ANK2 Mouse Monoclonal Antibody [clone: S105-17]
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: ANK2 (ankyrin 2, neuronal)
Clonality: monoclonal
Clone: S105-17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR