Anti-BCAN Mouse Monoclonal Antibody [clone: S294A-6]

Leverandør: US Biological
167578
USBI167578EA 7225 NOK
USBI167578
Anti-BCAN Mouse Monoclonal Antibody [clone: S294A-6]
Antistoffer
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: BCAN (brevican)
Clonality: monoclonal
Clone: S294A-6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR