Anti-Copper Transporting ATPase2 Mouse Monoclonal Antibody [clone: S62-29]

Leverandør: US Biological
167582
USBI167582EA 7225 NOK
USBI167582
Anti-Copper Transporting ATPase2 Mouse Monoclonal Antibody [clone: S62-29]
Antistoffer
Type: Primary
Antigen: Copper Transporting ATPase2
Clonality: monoclonal
Clone: S62-29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR