Anti-DICER1 Mouse Monoclonal Antibody [clone: S167-7]

Leverandør: US Biological
167583
USBI167583EA 7225 NOK
USBI167583
Anti-DICER1 Mouse Monoclonal Antibody [clone: S167-7]
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: DICER1 (dicer 1, ribonuclease type III)
Clonality: monoclonal
Clone: S167-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR