Anti-TXNDC5 Mouse Monoclonal Antibody [clone: 2E7/7]

Leverandør: US Biological
167585
USBI167585EA 7225 NOK
USBI167585
Anti-TXNDC5 Mouse Monoclonal Antibody [clone: 2E7/7]
Antistoffer
Recommended dilution: Western Blot: 1:200

Type: Primary
Antigen: TXNDC5 (thioredoxin domain containing 5 (endoplasmic reticulum))
Clonality: monoclonal
Clone: 2E7/7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR