Anti-BSA Mouse Monoclonal Antibody [clone: 14B4]

Leverandør: US Biological
168015
USBI168015EA 13275 NOK
USBI168015
Anti-BSA Mouse Monoclonal Antibody [clone: 14B4]
Antistoffer
Recommended dilution: ELISA: 1:120000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:100

Type: Primary
Antigen: BSA (Bovine Serum Albumin)
Clonality: monoclonal
Clone: 14B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR