Anti-Biotin Rabbit Polyclonal Antibody Rhodamine

Leverandør: US Biological
168058
USBI168058EA 9525 NOK
USBI168058
Anti-Biotin Rabbit Polyclonal Antibody Rhodamine
Antistoffer
Recommended dilution: Immunofluorescence: 1:500-1:2500

Type: Tag
Antigen: Biotin (biotin)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR