Anti-KLH Mouse Monoclonal Antibody [clone: 14B31]

Leverandør: US Biological
168322
USBI168322EA 8550 NOK
USBI168322
Anti-KLH Mouse Monoclonal Antibody [clone: 14B31]
Antistoffer
Recommended dilution: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000

Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: monoclonal
Clone: 14B31
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR