Anti-KLH Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: US Biological
168323
USBI168323EA 10275 NOK
USBI168323
Anti-KLH Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:15000-1:60000
Do not freeze!

Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR