Anti-KLH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: US Biological
168324
USBI168324EA 10275 NOK
USBI168324
Anti-KLH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:400000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500

Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR