Anti-KLH Rabbit Polyclonal Antibody Rhodamine

Leverandør: US Biological
168325
USBI168325EA 10375 NOK
USBI168325
Anti-KLH Rabbit Polyclonal Antibody Rhodamine
Antistoffer
Recommended dilution: Immunofluorescence: 1:500-1:2500

Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol KLH
Antigen-navn Keyhole Limpet Hemocyanin
Environmentally Preferable
Antistofftype Tag
Klonalitet Polyclonal
Konjugering TRITC (Tetramethylrhodamine Isothiocyanate)
Reaktivitet
Vert Rabbit
Isotype IgG
ImmunoFluorescens Yes
Skjema lyophilized powder
Rensing purified by immunoaffinity chromatography
Konsentrasjon ~1 mg/ml (after reconstitution)
Lagringsbuffer 0,02M PBS, pH 7,2, 0,01% sodium azide, 10mg/ml BSA.
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR