Anti-R-Phycoerythrin Goat Polyclonal Antibody Rhodamine

Leverandør: US Biological
168459
USBI168459EA 14025 NOK
USBI168459
Anti-R-Phycoerythrin Goat Polyclonal Antibody Rhodamine
Antistoffer
Recommended dilution: Immunofluorescence: 1:500-1:2500; Flow Cytometry: 1:2000-1:10000

Type: Tag
Antigen: R-Phycoerythrin
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR