Anti-Activation-induced Cytidine Deaminase Mouse Monoclonal Antibody [clone: 8k266]

Leverandør: US Biological
A0855-75H-100
USBIA0855-75H-100EA 5475 NOK
USBIA0855-75H-100
Anti-Activation-induced Cytidine Deaminase Mouse Monoclonal Antibody [clone: 8k266]
Antistoffer
Recommended dilution: ELISA: 1:1000-1:2000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Activation-induced Cytidine Deaminase
Clonality: Monoclonal
Clone: 8k266
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR